missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTR10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567160
This item is not returnable.
View return policy
Description
Immunogen sequence: KALGTKTFRIH TPPAKANCVA WLGGAIFGAL QDILGSRSVS KEYYNQTGRI PDWCSLNNPP LEMMFDVGKT QPPLMK Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
ACTR10is a protein coding gene. Gene Ontology (GO) annotations related to this gene include neutrophil degranulation.
Specifications
| ACTR10 | |
| Polyclonal | |
| Unconjugated | |
| ACTR10 | |
| Act11; actin related protein 10; actin related protein 10 homolog; actin-related protein 10; actin-related protein 10 homolog; actin-related protein 10 homolog (S. cerevisiae); actin-related protein 10-like protein; actin-related protein 11; Actr10; ACTR11; Arp10; ARP10 actin related protein 10 homolog; ARP10 actin-related protein 10; ARP10 actin-related protein 10 homolog; Arp11; hARP11; hypothetical protein LOC511083; LOW QUALITY PROTEIN: actin-related protein 10; zgc:55635 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55860 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZ32 | |
| ACTR10 | |
| Recombinant Human ACTR10. Recombinant protein control fragment (Product #RP-105526). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction