Learn More
ACTR3 Antibody (7E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26525-50ul
 View more versions of this product
Product Code. 30043642
  • € 343.00 / 50µL
Description
ACTR3 Monoclonal antibody specifically detects ACTR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACTR3
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA
7E5
Western Blot 1:500-1:5000, ELISA
actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3
A synthesized peptide derived from Human ACTR3 [UniProt P61158]
50 μL
Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity
10096
Store at -20 to -70C. Avoid freeze-thaw cycles.