missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTRT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00140625-D01P
This item is not returnable.
View return policy
Description
ACTRT2 Polyclonal antibody specifically detects ACTRT2 in Human samples. It is validated for Western Blot
Specifications
| ACTRT2 | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_536356.3 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 140625 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| actin-related protein hArpM2, Actin-related protein M2, actin-related protein T2, ARPM2HARPM2, Arp-T2, ARPT2, FLJ25424 | |
| ACTRT2 (NP_536356.3, 1 a.a. - 377 a.a.) full-length human protein. MFNPHALDSPAVIFDNGSGFCKAGLSGEFGPRHMVSSIVGHLKFQAPSAEANQKKYFVGEEALYKQEALQLHSPFERGLITGWDDVERLWKHLFEWELGVKPSDQPLLATEPSLNPRENREKMAEVMFENFGVPAFYLSDQAVLALYASACVTGLVVDSGDAVTCTVPIFEGYSLPHAVTKLHVAGRDITELLMQLLLASGHTFPCQLDKGLVDDIKKKLCYVALEPEKELSRRPEEVLREYKLPDGNIISLGDPLHQAPEALFVPQQLGSQSPGLSNMVSSSITKCDTDIQKILFGEIVLSGGTTLFHGLDDRLLKELEQLASKDTPIKITAPPDRWFSTWIGASIVTSLSSFKQMWVTAADFKEFGTSVVQRRCF | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction