missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTRT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555228
This item is not returnable.
View return policy
Description
Immunogen sequence: ALEPEKELSR RPEEVLREYK LPDGNIISLG DPLHQAPEAL FVPQQLGSQS PGLSNMVSSS ITKCDTDIQK ILFGEIVLSG GTTLFHGLDD RLLKELEQLA SKDTPIKITA P Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.
The protein encoded by this intronless gene belongs to the actin family. Studies have shown that this protein may be involved in cytoskeletal organization similar to other cytoplasmic actin-related protein (ARP) subfamily members. Antibody raised against the human protein has been used to detect the protein by immunoblotting and immunofluorescence microscopy, demonstrating its specific synthesis in the testis, late in spermatid differentiation, and its localization in the calyx.
Specifications
| ACTRT2 | |
| Polyclonal | |
| Unconjugated | |
| ACTRT2 | |
| 1700052K15Rik; actin related protein M2 (Arpm2); actin related protein T2; actin-related protein hArpM2; actin-related protein M2; Actin-related protein T2; Actrt2; ARPM2; ARPT2; ARP-T2; HARPM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDY3 | |
| ACTRT2 | |
| Recombinant protein corresponding to Human ACTRT2. Recombinant protein control fragment (Product #RP-88853). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction