Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Product Code. 10254312
Click to view available options
Volume (Metric):
0.5-10 μL, 10-100 μL, 100-1000 μL
10, 100, 1000 μL
10, 50, 200 μL
10-100 μL, 100-1000 μL, 0.5-5.0 mL
100-1000 μL, 0.5-5.0, 1.0-10 mL
2, 10, 50 μL
2, 20, 200 μL
2-20 μL, 20-200 μL, 100-1000 μL
5-50 μL, 20-200 μL, 100-1000 μL
Unit Size:
1 set
Product Code. 10254312
Brand: Socorex™ 835.TRIO.Y
Only null left
Add to basket
Add to basket
Classic and universal, these micropipettes are intended for all applications and come in a select three-pack. Socorex™ Acura™ Manual 825 Micropipette Triopack micropipettes are designed for comfort and sturdiness.
  • Ergonomic shape.
  • Lightweight.
  • Uultra-soft plunger stroke.
  • Smart and reliable volume adjustment:
    • State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
    • Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
  • Digital display of set volume on the front of the instrument is easily visible during pipetting.
  • Autoclavable.
Specifications
Certifications/Compliance EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices
Autoclavable Autoclavable
Product Line Acura
Type Micropipette Triopack
Volume (Metric) 100-1000 μL, 0.5-5.0, 1.0-10 mL
Includes 3 Acura™ micropipettes, manual, spike samples, Qualitips™ 1000μL/5mL/10mL
Quantity 3/Pk.