Learn More
ACVRL1 Monoclonal Antibody (OTI5C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809560
Encompass_Preferred
 View more versions of this product
Product Code. 17501944
  • € 540.87 / 100µL
Description
The TGF-beta superfamily of ligands exerts its biological activity by binding to heteromeric receptor complexes two types (I and II) of the serine/threonine kinases. ALK1, like the other type I receptors, contains a cysteine rich domain with conserved cysteine spacing in the extracellular region and a glycine and serine rich domain preceding the kinase domain. The reduced human ALK1 monomer has a calculated molecular mass of approximately 37.3 kDa. Due to glycoslyation, the recombit human protein migrates as a 50-55 kDa protein in SDS-PAGE under reducing conditions. Human and mouse ALK1 share approximately 71% amino acid sequence identity in their extracellular domains. ALK1 is highly expressed in endothelial cells and other vascular tissues.
Specifications
Specifications
ACVRL1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P37023
ACVRL1
Human recombit protein fragment corresponding to amino acids 22-119 of human ACVRL1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI5C2
Unconjugated
ACVRL1
ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I
Mouse
Affinity Chromatography
RUO
94
-20° C, Avoid Freeze/Thaw Cycles