Learn More
ACVRL1 Monoclonal Antibody (OTI5C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809560S
This item is not returnable.
View return policy
Description
ACVRL1 Monoclonal antibody specifically detects ACVRL1 in Human samples. It is validated for Western BlotSpecifications
| ACVRL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01160 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4878 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5C2 | |
| Unconjugated | |
| NPPA | |
| activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I | |
| Human recombinant protein fragment corresponding to amino acids 26-151 of human NPPA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |