Learn More
ACVRL1 Monoclonal Antibody (OTI5C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809560S
Encompass_Preferred
 View more versions of this product
Product Code. 17521944
  • € 242.00 / 30µL
Description
ACVRL1 Monoclonal antibody specifically detects ACVRL1 in Human samples. It is validated for Western Blot
Specifications
Specifications
ACVRL1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P01160
Mouse
Affinity chromatography
RUO
4878
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5C2
Unconjugated
NPPA
activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I
Human recombinant protein fragment corresponding to amino acids 26-151 of human NPPA produced in E.coli.
30 μL