missing translation for 'onlineSavingsMsg'
Learn More

ACY-775, MedChemExpress

Product Code. 16447596
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447596

Brand: Medchem Express HY19328

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ACY-775 is a potent and selective inhibitor of the of histone deacetylase 6 (HDAC6) with an IC50 of 7.5 nM. ACY775 also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

ACY-775 is a potent and selective inhibitor of the of histone deacetylase 6 (HDAC6) with an IC50 of 7.5 nM. ACY775 also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Specifications

Chemical Name or Material ACY-775
CAS 1375466-18-4
Color White
Purity Grade Notes Research
Molecular Formula C17H19FN4O2
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (75.68 mM; Need ultrasonic)
SMILES O=C(NO)C(C=N1)=CN=C1NC2(CCCCC2)C3=CC=CC(F)=C3
Molecular Weight (g/mol) 330.36
Formula Weight 330.36
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.