Learn More
Invitrogen™ ACY3 Monoclonal Antibody (OTI4C1)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525390
Product Code. 15944994
  • € 588.00 / 100µL
Description
ACY3 Monoclonal Antibody for Western Blot, IHC (P), Flow
ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACY3
Monoclonal
0.41 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96HD9
Acy3
Full length human recombinant protein of ACY3 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI4C1
Unconjugated
Acy3
0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394
Mouse
Affinity Chromatography
RUO
91703
-20°C, Avoid Freeze/Thaw Cycles