missing translation for 'onlineSavingsMsg'
Learn More

Acyclovir N-Ethyl-L-valinate, TRC

Product Code. 30326906
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326906 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326906 Supplier Toronto Research Chemicals Supplier No. TRCA61860550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-Ethyl-L-valinate
CAS 1346747-69-0
Molecular Formula C15 H24 N6 O4
Quantity 50 mg
Synonym L-Valine, N-ethyl-, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester, Valaciclovir Hydrochloride Imp. D (EP), Valacyclovir RC D (USP), USP Valacyclovir Related Compound D, GW388148, Ph Eur Valaciclovir Impurity D
InChI Formula InChI=1S/C15H24N6O4/c1-4-17-10(9(2)3)14(23)25-6-5-24-8-21-7-18-11-12(21)19-15(16)20-13(11)22/h7,9-10,17H,4-6,8H2,1-3H3,(H3,16,19,20,22)/t10-/m0/s1
SMILES CCN[C@@H](C(C)C)C(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N
IUPAC Name 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl (2S)-2-(ethylamino)-3-methylbutanoate
Molecular Weight (g/mol) 352.39
Formula Weight 352.1859
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.