missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acylglycerol Kinase Antibody (9A3B1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 468.00
Specifications
| Antigen | Acylglycerol Kinase |
|---|---|
| Clone | 9A3B1 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629386
|
Novus Biologicals
NBP3-05339-100ul |
100 μg |
€ 468.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630237
|
Novus Biologicals
NBP3-05339-20ul |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acylglycerol Kinase Monoclonal antibody specifically detects Acylglycerol Kinase in Human samples. It is validated for Western BlotSpecifications
| Acylglycerol Kinase | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| acylglycerol kinase, EC 2.7.1.107, EC 2.7.1.94, FLJ10842, hAGK, HsMuLK, mitochondrial, MuLK, MULKhsMuLK, Multi-substrate lipid kinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-422 of human Acylglycerol Kinase (NP_060708.1). MTVFFKTLRNHWKKTTAGLCLLTWGGHWLYGKHCDNLLRRAACQEAQVFGNQLIPPNAQVKKATVFLNPAACKGKARTLFEKNAAPILHLSGMDVTIVKTDYEGQAKKLLELMENTDVIIVAGGDGTLQEVVTGVLRRTDEATFSKIPIGFIPLGETSSLSHTLFAESGNKVQHITDATLAIVKGETVPLDVLQIKGEKEQPVFAMTGLRWGSFRDAGVKVSKYWYLGPLKIKAAHFFSTLKEWPQTHQASISYTGPTERPPNEPEETPVQRPSLYRRILRRLASYWAQPQDALSQEVSPEVWKDVQLSTIELSITTRNNQLDPTSKEDFLNICIEPDTISKGDFITIGSRKVRNPKLHVEGTECLQASQCTLLIPEGAGGSFSIDSEEYEAMPVEVKLLPRKLQFFCDPRKREQMLTSPTQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9A3B1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 55750 | |
| IgG1 κ | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title