Learn More
ACYP1 Antibody (004), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-90125CL1
Product Code. 30143008
  • € 1020.00 / 0.10mL
Description
ACYP1 Monoclonal antibody specifically detects ACYP1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACYP1
Monoclonal
CoraFluor 1
acylphosphatase 1, erythrocyte (common) type, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1, ACYPEacylphosphatase-1, EC 3.6.1.7
This antibody was obtained from a rabbit immunized with purified, recombinant Human ACYP1 (Uniprot#: P07311; Ala 2-Lys 99).
0.1 mL
Cancer, Endocrinology, Signal Transduction
97
Store at 4°C in the dark. Do not freeze.
IgG
ELISA
004
PBS
Rabbit
Protein A purified