Learn More
ACYP1 Antibody (004), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90125PCP
Product Code. 18631832
  • € 607.00 / 0.10mL
Description
ACYP1 Monoclonal antibody specifically detects ACYP1 in Human samples. It is validated for ELISA