missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACYP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518639
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Acylphosphatase is a small cytosolic enzyme that catalyzes the hydrolysis of the carboxyl-phosphate bond of acylphosphates. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localization. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis.
Specifications
| ACYP1 | |
| Polyclonal | |
| Unconjugated | |
| ACYP1 | |
| 1110039O14Rik; acylphosphatase 1; acylphosphatase 1, erythrocyte (common) type; acylphosphatase, erythrocyte isozyme; Acylphosphatase, organ-common type isozyme; acylphosphatase-1; Acylphosphate phosphohydrolase 1; Acyp1; ACYPE; AI325944 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 97 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P07311 | |
| ACYP1 | |
| Synthetic peptide sequence (PKSHIDKANFNNEK) corresponding to the internal amino acids of ACYP1 (aa 72-86). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction