missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADA2 beta Antibody (3F3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00093624-M06
This item is not returnable.
View return policy
Description
ADA2 beta Monoclonal antibody specifically detects ADA2 beta in Human samples. It is validated for Western Blot, ELISA
Specifications
| ADA2 beta | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 93624 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 3F3 | |
| Western Blot 1:500, ELISA 1:100-1:2000 | |
| ADA2-beta, ADA2-like protein beta, MGC21874, transcriptional adapter 2-beta, transcriptional adaptor 2 (ADA2 homolog, yeast)-beta, transcriptional adaptor 2B | |
| MGC21874 (XP_291105, 2 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AELGKKYCVYCLAEVSPLRFRCTECQDIELCPECFSAGAEIGHHRRYHGYQLVDGGRFTLWGPEAEGGWTSREEQLLLDAIEQFGFGNWEDMAAHVGASRTPQEVMEHY | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction