missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21217-1-AP-150UL
This item is not returnable.
View return policy
Description
ADAD2 is a protein coding gene.Specifications
| ADAD2 | |
| Polyclonal | |
| Unconjugated | |
| ADAD2 | |
| ADAD2, TENRL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 161931, 691275, 75773 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NCV1, Q9D5P4 | |
| Adad2 | |
| ADAD2 Fusion Protein Ag15650 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction