missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Antibody (139712), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB946CL1
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal antibody specifically detects ADAM10 in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| ADAM10 | |
| Monoclonal | |
| CoraFluor 1 | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM10, Thr215-Glu673, Accession # O35598 | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 102 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Western Blot, CyTOF | |
| 139712 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction