missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Antibody (MM0077-6D31), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12014AF750
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ADAM10 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ADAM10 | |
| Human recombinant ADAM10 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| In Western blots, this antibody shows weak cross-reactivity with recombinant mouse ADAM10 and human ADAM8, 9, 15, and 17. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0077-6D31 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Signal Transduction | |
| 102.0 | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction