missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM10 Antibody (RM0146-7H12), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12015V
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal specifically detects ADAM10 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| ADAM10 | |
| Monoclonal | |
| DyLight 405 | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Detects mouse ADAM10 and cross reacts with human ADAM10. Weak cross-reactivity is observed to ADAM9 and ADAM15 in Western blotting. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| RM0146-7H12 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| ADAM10 | |
| Mouse recombinant ADAM10 | |
| 0.1 mL | |
| Apoptosis, Signal Transduction | |
| 102 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction