missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM15 Antibody (001), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90765AF647
This item is not returnable.
View return policy
Description
ADAM15 Monoclonal specifically detects ADAM15 in Mouse samples. It is validated for ELISA.
Specifications
| ADAM15 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ADAM15 (O88839-1; Met1-Thr697). | |
| 0.1 mL | |
| Primary | |
| Has cross-reactivity in ELISA with: Mouse ADAM9 No cross-reactivity in ELISA with: Human ADAM15 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 1 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8751 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction