missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM15 Antibody (23G9) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB935JF635
This item is not returnable.
View return policy
Description
ADAM15 Monoclonal antibody specifically detects ADAM15 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| ADAM15 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant human ADAM15, Asp207-Thr696, Accession # Q13444 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 23G9 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 8751 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction