missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM15 Antibody (MM0078-9H34), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12017AF594
This item is not returnable.
View return policy
Description
ADAM15 Monoclonal specifically detects ADAM15 in Human samples. It is validated for Western Blot.
Specifications
| ADAM15 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ADAM15 | |
| Human recombinant ADAM15 (extracellular domain) | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| MM0078-9H34 | |
| Western Blot, Immunohistochemistry | |
| ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
| Mouse | |
| 0.1 mL | |
| 8751 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction