missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM15 Antibody (SR2269), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22227-50ul
This item is not returnable.
View return policy
Description
ADAM15 Monoclonal antibody specifically detects ADAM15 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ADAM15 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2269 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
| A synthesized peptide derived from human ADAM15 (Uniprot #: Q13444) | |
| 50 μg | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 8751 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction