missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM15 Recombinant Rabbit Monoclonal Antibody (4T4X10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535694
This item is not returnable.
View return policy
Description
Immunogen sequence: RHIRRRRDVV TETKTVELVI VADHSEAQKY RDFQHLLNRT LEVALLLDTF FRPLNVRVAL VGLEAWTQRD LVEISPNPAV TLENFLHWRR AHLLPRLPHD S.
The protein encoded by this gene is a member of the ADAM (a disintegrin and metalloproteinase) protein family. ADAM family members are type I transmembrane glycoproteins known to be involved in cell adhesion and proteolytic ectodomain processing of cytokines and adhesion molecules. This protein contains multiple functional domains including a zinc-binding metalloprotease domain, a disintegrin-like domain, as well as a EGF-like domain. Through its disintegrin-like domain, this protein specifically interacts with the integrin beta chain, beta 3. It also interacts with Src family protein-tyrosine kinases in a phosphorylation-dependent manner, suggesting that this protein may function in cell-cell adhesion as well as in cellular signaling. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| ADAM15 | |
| Recombinant Monoclonal | |
| 3 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88839, Q13444, Q9QYV0 | |
| Adam15 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4T4X10 | |
| Unconjugated | |
| Adam15 | |
| a disintegrin and metallopeptidase domain 15 (metargidin); a disintegrin and metalloprotease domain 15 (metargidin); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 15; a disintegrin and metalloproteinase domain 15 (metargidin); AD56; ADAM; ADAM 15; ADAM metallopeptidase domain 15; Adam15; ADAMs; CRII-7; disintegrin and metalloprotease 15; disintegrin and metalloproteinase domain-containing protein 15; Mdc15; MDC-15; metalloendopeptidases; Metalloprotease RGD disintegrin protein; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 15; metargidin; tMDC VI; tMDCVI | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11490, 57025, 8751 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction