missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAM19 Polyclonal Antibody

Product Code. 13291109
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13291109 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13291109 Supplier Invitrogen™ Supplier No. PA526816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADAM19 Polyclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a type I transmembrane protein and serves as a marker for dendritic cell differentiation. It has also been demonstrated to be an active metalloproteinase, which may be involved in normal physiological and pathological processes such as cells migration, cell adhesion, cell-cell and cell-matrix interactions, and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM19
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADAM19
Gene Accession No. Q9H013
Gene Alias a disintegrin and metallopeptidase domain 19 (meltrin beta); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 19 (meltrin beta); ADAM; ADAM 19; ADAM metallopeptidase domain 19; ADAM19; ADAMs; AL024287; disintegrin and metalloproteinase domain-containing protein 19; FKSG34; MADDAM; meltrin beta; meltrin-beta; metalloendopeptidases; Metalloprotease and disintegrin dendritic antigen marker; metalloprotease-disintegrin meltrin beta; MLTNB; Sox30; SRY-box containing gene 30
Gene Symbols ADAM19
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 207-236 amino acids from the central region of human ADAM19.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8728
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.