missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAM21 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15664235
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15664235 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15664235 Supplier Invitrogen™ Supplier No. PA550305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ADAM21 protein.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The expression of this gene expression is testis-specific.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM21
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ADAM21
Gene Accession No. Q9UKJ8
Gene Alias a disintegrin and metallopeptidase domain 21; a disintegrin and metalloprotease domain (ADAM) 21; a disintegrin and metalloprotease domain 21; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 21; ADAM; ADAM 21; ADAM 31; ADAM metallopeptidase domain 21; ADAM metallopeptidase domain 21, preproprotein; Adam21; Adam31; ADAMs; disintegrin and metalloproteinase domain-containing protein 21; Disintegrin and metalloproteinase domain-containing protein 31; metalloendopeptidases
Gene Symbols ADAM21
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human ADAM metallopeptidase domain 21.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8747
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.