missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ADAM22 Polyclonal specifically detects ADAM22 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ADAM22 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q9R1V6 |
| Gene Alias | ADAM 22, ADAM metallopeptidase domain 22, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832 |
| Gene Symbols | ADAM22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?