missing translation for 'onlineSavingsMsg'
Learn More

ADAM22 Antibody, Novus Biologicals™

Product Code. p-7108648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18281022 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18281022 Supplier Novus Biologicals Supplier No. NBP169015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADAM22 Polyclonal specifically detects ADAM22 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM22
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9R1V6
Gene Alias ADAM 22, ADAM metallopeptidase domain 22, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832
Gene Symbols ADAM22
Host Species Rabbit
Immunogen Synthetic peptides corresponding to Adam22 (a disintegrin and metallopeptidase domain 22) The peptide sequence was selected from the N terminal of mouse Adam22 (NP_001007222). Peptide sequence AISENPLITLREFMKYRRDFIKEKADAVHLFSGSQFESSRSGAAYIGGIC.
Molecular Weight of Antigen 91 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53616
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.