missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM23 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98071JF549
This item is not returnable.
View return policy
Description
ADAM23 Polyclonal specifically detects ADAM23 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ADAM23 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| a disintegrin and metalloproteinase domain 23, ADAM metallopeptidase domain 23, disintegrin and metalloproteinase domain-containing protein 23, MDC-3, MDC3ADAM 23, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human ADAM23 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8745 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction