missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98071-100ul
This item is not returnable.
View return policy
Description
ADAM23 Polyclonal antibody specifically detects ADAM23 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ADAM23 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| a disintegrin and metalloproteinase domain 23, ADAM metallopeptidase domain 23, disintegrin and metalloproteinase domain-containing protein 23, MDC-3, MDC3ADAM 23, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human ADAM23 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8745 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?