missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM33 Antibody (508101), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5565D
This item is not returnable.
View return policy
Description
ADAM33 Monoclonal antibody specifically detects ADAM33 in Human samples. It is validated for Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| ADAM33 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF | |
| 508101 | |
| Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM33, Val30-Leu501 (predicted), Accession # Q9BZ11 | |
| 0.1 mL | |
| Asthma, Cell Cycle and Replication, Immunology | |
| 80332 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction