missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAM7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15740571
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15740571 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15740571 Supplier Invitrogen™ Supplier No. PA552620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LGVEGQQLVR PKKLPLIQKR DTGHTHDDDI LKTYEEELLY EIKLNRKTLV LHLLRSREFL GSNYSETFYS MKGEAFTRHP QIMDHCFYQG SIVHEYDSAA SISTCNGLRG FFRINDQRYL IEPVKYSDEG EHLVFKYNLR VPYGANYS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.

The ADAM family is composed of zinc-binding proteins that can function as adhesion proteins and/or endopeptidases. They are involved in a number of biologic processes, including fertilization, neurogenesis, muscle development, and immune response.The ADAM family is composed of zinc-binding proteins that can function as adhesion proteins and/or endopeptidases. They are involved in a number of biologic processes, including fertilization, neurogenesis, muscle development, and immune response.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAM7
Gene Accession No. Q9H2U9
Gene Alias 9230104J12Rik; a disintegrin and metallopeptidase domain 7; a disintegrin and metalloprotease domain (ADAM) 7; a disintegrin and metalloprotease domain 7; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 7; ADAM; ADAM 7; ADAM metallopeptidase domain 7; Adam7; ADAM-7; ADAMs; Disintegrin and metalloproteinase domain-containing protein 7; EAP1; EAPI; epididymal apical protein I; GP83; GP-83; metalloendopeptidases; Sperm maturation-related glycoprotein GP-83
Gene Symbols ADAM7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAM7. Recombinant protein control fragment (Product #RP-89230).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8756
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.