missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Antibody (143338) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10311JF635
This item is not returnable.
View return policy
Description
ADAM8 Monoclonal antibody specifically detects ADAM8 in Human samples. It is validated for Immunoprecipitation,Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| ADAM8 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain, Asp158-Ser653, Accession # P78325 | |
| 0.5 mL | |
| Cellular Markers | |
| 101 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunoprecipitation, Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 143338 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction