missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Antibody (MM0079-6J12), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12020UV
This item is not returnable.
View return policy
Description
ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot.
Specifications
| ADAM8 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ADAM8 | |
| Human recombinant ADAM8 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0079-6J12 | |
| Western Blot | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Mouse | |
| Protein G purified | |
| Cellular Markers | |
| 101 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction