missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM9 Antibody (MM0080-8A15), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12021AF594
This item is not returnable.
View return policy
Description
ADAM9 Monoclonal specifically detects ADAM9 in Human samples. It is validated for Western Blot.
Specifications
| ADAM9 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ADAM9 | |
| Human recombinant ADAM9 (extracellular domain) | |
| Primary | |
| Detects human ADAM9 with low cross activity with mouse ADAM9 | |
| Store at 4°C in the dark. |
| Western Blot | |
| MM0080-8A15 | |
| Western Blot | |
| a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
| Mouse | |
| 0.1 mL | |
| 8754 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction