missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99535-100ul
This item is not returnable.
View return policy
Description
ADAMTS10 Polyclonal antibody specifically detects ADAMTS10 in Mouse samples. It is validated for Western Blot
Specifications
| ADAMTS10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 10, ADAM metallopeptidase with thrombospondin type 1 motif, 10, ADAM-TS 10, ADAMTS-10, ADAM-TS10A disintegrin and metalloproteinase with thrombospondin motifs 10, EC 3.4.24.-, EC 3.4.24.82, WMS, zinc metalloendopeptidase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ADAMTS10 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 81794 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction