missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS13 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-55261AF532
This item is not returnable.
View return policy
Description
ADAMTS13 Polyclonal antibody specifically detects ADAMTS13 in Human samples. It is validated for Western Blot
Specifications
| ADAMTS13 | |
| Polyclonal | |
| Western Blot | |
| A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTPADAM-TS 13, vWF-cleaving protease, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease | |
| The immunogen recognized by this antibody maps to a region between residue 825 and 875 of human ADAM (a disintegrin-like and metalloprotease) metallopeptidase with thrombospondin type 1 motif, 13 using the numbering given in entry NP_620594.1 (GeneID 1109 | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 11093 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction