missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS15 Antibody (561819), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5149PECY55
This item is not returnable.
View return policy
Description
ADAMTS15 Monoclonal antibody specifically detects ADAMTS15 in Human samples. It is validated for Flow Cytometry
Specifications
| ADAMTS15 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 170689 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 561819 | |
| Flow Cytometry | |
| A disintegrin and metalloproteinase with thrombospondin motifs 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, preproprotein, ADAM metallopeptidase with thrombospondin type 1 motif, 15, ADAM-TS 15, ADAM-TS15, ADAMTS-15, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, metalloprotease disintegrin 15 with thrombospondin domains, MGC126403 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS15, Gly18-Cys682, Accession # Q8TE58 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction