missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTS20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555665
This item is not returnable.
View return policy
Description
Immunogen sequence: STRPCSQRRC WSQDCVQHKG MERGRLNCST SCERKDSHQR MECTDNQIRQ VNEIVYNSST ISLTSKNCRN PPCNYIVVTA DSSQCANN Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 45%.
May play a role in tissue-remodeling process occurring in both normal and pathological conditions. May have a protease-independent function in the transport from the endoplasmic reticulum to the Golgi apparatus of secretory cargos, mediated by the GON domain.
Specifications
| ADAMTS20 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTS20 | |
| a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 20; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 20; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 20; ADAM metallopeptidase with thrombospondin type 1 motif, 20; ADAMs; ADAM-TS 20; ADAMTS20; ADAM-TS20; ADAMTS-20; GON-1; metalloendopeptidases | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80070 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P59510 | |
| ADAMTS20 | |
| Recombinant protein corresponding to Human ADAMTS20. Recombinant protein control fragment (Product #RP-95142). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction