missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS4 Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805430S
This item is not returnable.
View return policy
Description
ADAMTS4 Monoclonal antibody specifically detects ADAMTS4 in Human samples. It is validated for Western BlotSpecifications
| ADAMTS4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99801 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4824 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| NKX3-1 | |
| 3830423K05; a disintegrin and metallopeptidase with thrombospondin motifs 4; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 4; ADAM metallopeptidase with thrombospondin type 1 motif, 4; ADAMs; ADAM-TS 4; ADAMTS family member; ADAMTS-2; Adamts4; ADAM-TS4; ADAMTS-4; ADMP-1; Aggrecanase; Aggrecanase 1; aggrecanase-1; disintegrin and metalloproteinase with thrombospondin motifs 2; KIAA0688; metalloendopeptidases; metalloproteinase; mKIAA0688; UNQ769/PRO1563 | |
| Full length human recombinant protein of human NKX3-1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction