Learn More
ADAMTS4 Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA805430
Encompass_Preferred
 View more versions of this product
Product Code. 17535353
  • € 540.87 / 100µL
Description
This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene lacks a C-terminal TS motif. It is responsible for the degradation of aggrecan, a major proteoglycan of cartilage, and brevican, a brain-specific extracellular matrix protein. The cleavage of aggrecan and brevican suggests key roles of this enzyme in arthritic disease and in the central nervous system, potentially, in the progression of glioma.
Specifications
Specifications
ADAMTS4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75173
ADAMTS4
Human recombit protein fragment corresponding to amino acids 296-577 of human ADAMTS4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2G8
Unconjugated
ADAMTS4
ADAMTS-2, ADAMTS-4, ADMP-1
Mouse
Affinity Chromatography
RUO
9507
-20° C, Avoid Freeze/Thaw Cycles