Learn More
ADAMTSL-1/Punctin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-92531-0.02ml
 Afficher plus de versions de ce produit
Code produit. 18609771
  • € 190.00 / 0.02mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
ADAMTSL-1/Punctin Polyclonal antibody specifically detects ADAMTSL-1/Punctin in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spécification
Spécification
ADAMTSL-1/Punctin
Polyclonal
Western Blot 1:500-1:2000
ADAM-TS related protein 1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1
Recombinant fusion protein containing a sequence corresponding to amino acids 1483-1762 of human ADAMTSL1 (NP_001035362.3). SLVIQDYWWSVDRLATCSASCGNRGVQQPRLRCLLNSTEVNPAHCAGKVRPAVQPIACNRRDCPSRWMVTSWSACTRSCGGGVQTRRVTCQKLKASGISTPVSNDMCTQVAKRPVDTQACNQQLCVEWAFSSWGQCNGPCIGPHLAVQHRQVFCQTRDGITLPSEQCSALPRPVSTQNCWSEACSVHWRVSLWTLCTATCGNYGFQSRRVECVHARTNKAVPEHLCSWGPRPANWQRCNITPCENMECRDTTRYCEKVKQLKLCQLSQFKSRCCGTCGKA
0.02 mL
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
92949
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS with 50% glycerol, pH7.3.