missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTSL1 Monoclonal Antibody (1B2-8-2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533368
This item is not returnable.
View return policy
Description
Product is supplied at 1 mL (100 μL/mL). Western blotting applications the antibody works better in non-reducing than reducing conditions although it will also recognize the reduced antigen at correspondingly higher concentrations.
Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
Specifications
| ADAMTSL1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Q8N6G6 | |
| ADAMTSL1 | |
| Protein G | |
| RUO | |
| 92949 | |
| 4°C | |
| Liquid |
| ELISA, Western Blot | |
| 1B2-8-2 | |
| Unconjugated | |
| ADAMTSL1 | |
| 5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction