Learn More
Invitrogen™ ADAMTSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139930
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: REHFVIKLIG GNRKLVARPL SPRSEEEVLA GRKGGPKEAL QTHKHQNGIF SNGSKAEKRG LAANPGSRYD DLVSRLL Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 83%.
Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
Specifications
| ADAMTSL1 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTSL1 | |
| 5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N6G6 | |
| ADAMTSL1 | |
| Recombinant protein corresponding to Human ADAMTSL1. Recombinant protein control fragment (Product #RP-109445). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.