Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 6453
Additional Details : Weight : 0.27000kg
 View more versions of this product
Product Code. 10025661
  • € 138.86 / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Adapter
Specifications
Specifications
Thread Size: M30 x 1.5 female to G 1 female
Adapter
16 x 1 m