Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Socket
Product Code. 10772522
Click to view available options
Standard Taper Joint No:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 25/29
Unit Size:
Pack of 10
Product Code. 10772522
Brand: Lenz Laborglasintrumente 01100112
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Sockets
  • Acc. to DIN 12249 form E
  • DIN standard
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Standard Taper Joint No NS 12/21
Thickness (Metric) Wall 1.8 mm
Diameter (Metric) Outer Tube 15 mm
Material Duran Glass