Description
- Adapter
- For 1 cryo containers (max. dia.: 13mm) or lidded centrifugation tubes (dia.: 12.2mm), max. length 50mm
- For Rotor F-45-18-17-Cryo
- Set of 6 pieces
Specifications
Specifications
| Type | Adapter |
| Tube Type | Cryotube |
| Capacity | 1 to 2 mL |
| For Use With (Equipment) | Eppendorf Angular Rotor |
| Bore Size (Metric) | 13 mm |
| No. of Places | 1 |
| No. per Pack | 6 Pack |