Learn More
IBA Lifesciences Adapter Set
Adapters for H-PR cartridges
Brand: IBA Lifesciences 2-1025-001
Additional Details : Weight : 0.20000kg
Description
TheStrep-tag™ purification system is based on the highly selective and easily controllable interaction between theStrep-tag™II peptide and the biotin binding site of a specially engineered streptavidin calledStrep-Tactin™.Strep-tag™II bindsStrep-Tactin™ nearly 100 times tighter than streptavidin, but elutes under gentle, physiological conditions.- Rapid one-step purification under physiological conditions
- Unsurpassed purity and bioactivity
Specifications
Adapter Set | |
it contains 6 x 1/16 Barb to 10-32 Male | |
For use with peristaltic pump tubing |
1 Ea. | |
Column Accesory |