Learn More
SI Analytics™ Conversion Adapter
Shop All SI Analytics Products
Click to view available options
For Use With (Application):
Conversion of all pH-meters with DIN entrance to BNC
Unit Size:
Each
Description
- Adapter for DIN socket
- Converts socket in BNC plug
- For conversion of all pH-meters with DIN entrance into BNC
Specifications
Specifications
| Description | Conversion Adapter |
| Type | Adapter DIN socket or BNC Connector |
| For Use With (Application) | Conversion of all pH-meters with DIN entrance to BNC |