Learn More
Eppendorf™ Adapters for Blood Tubes
Product Code. 11863422 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
2.6 to 7 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11863422 2.6 to 7 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11863422 Supplier Eppendorf™ Supplier No. 5804738004
Only null left
Add to basket
Add to basket
Add flexibility to Rotor F-34-6-38 for Eppendorf Centrifuge 5804/5804 R with these adapters
  • For use with Rotor F-34-6-38 and Eppendorf Centrifuge 5804/5804 R
  • For 3 round bottom tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Round Bottom
Capacity 2.6 to 7 mL
For Use With (Equipment) Eppendorf™ Rotor F-34-6-38
Bore Size (Metric) 13 mm
No. of Places 18
No. per Pack 2 Pack